Skip to Content
Merck
All Photos(1)

Key Documents

135259

Sigma-Aldrich

(3-Bromopropyl)triphenylphosphonium bromide

98%

Synonym(s):

NSC 84074

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)3P(C6H5)3Br
CAS Number:
Molecular Weight:
466.19
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

mp

228-230 °C (lit.)

functional group

bromo
phosphine

SMILES string

[Br-].BrCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C21H21BrP.BrH/c22-17-10-18-23(19-11-4-1-5-12-19,20-13-6-2-7-14-20)21-15-8-3-9-16-21;/h1-9,11-16H,10,17-18H2;1H/q+1;/p-1

InChI key

ZAHUZZUGJRPGKW-UHFFFAOYSA-M

Application

Reactant involved in:
  • Synthesis of functionalized polyurethanes using cationic ring-opening polymerization and click chemistry
  • Semipinacol rearrangement and direct arylation
  • Olefination of benzaldehydes
  • C-H activation / cycloisomerization
  • Intramolecular dehydrobromination
  • Cycloisomerizations of bromodienes and enynes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E W Czerwinski et al.
Acta crystallographica. Section C, Crystal structure communications, 44 ( Pt 10), 1862-1864 (1988-10-15)
C21H21BrP+.Br-, Mr = 464.2, monoclinic, P2(1)/c, a = 11.165(2), b = 10.160(2), c = 17.614(3)A, beta = 104.99(1) degree, V = 1930.08 A3, Z = 4, Dx = 1.597 g cm-3, graphite-monochromatized Cu K alpha radiation, lambda = 1.5418 A

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service