Documentos clave
58075
Tapered Flask for Filtration Apparatus 2
volume 2000 mL
About This Item
UNSPSC Code:
41115700
capacity
2000 mL
volume
2000 mL
General description
Tapered Flask for Filtration Apparatus 2 (volume 2000 mL) protects the instrument and columns by removing particles and gases from solvents and other mobile phase components.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Lot/Batch Number
Lo sentimos, en este momento no disponemos de COAs para este producto en línea.
Visite la Librería de documentos
Artículos
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.