Key Documents
55081
Frits for Upchurch Precolumn Filter
pore size 2.0 μm, pkg of 10 ea
About This Item
UNSPSC Code:
41115700
material
PEEK
packaging
pkg of 10 ea
pore size
2.0 μm
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
replacement part
Referencia del producto
Descripción
Precios
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Lot/Batch Number
Lo sentimos, en este momento no disponemos de COAs para este producto en línea.
Visite la Librería de documentos
Artículos
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.