Z742390
PELCO® Sliding Latch Microslide Storage Boxes
white, Holds 100 x slides 76x26 mm
Sinónimos:
slide storage
About This Item
Productos recomendados
material
acrylonitrile-butadiene-styrene (ABS)
white
feature
Holds 100 x slides 76x26 mm
packaging
pack of 12 ea
manufacturer/tradename
Ted Pella 2302-1-CS
L × W × H
209 mm (8.23 in.) × 172 mm (6.77 in.) × 32 mm (1.26 in.)
1 of 4
Este artículo | Z742398 | Z742396 | Z742405 |
---|---|---|---|
Holds 100 x slides 76x26 mm | Holds 50 x slides 76x26 mm | Holds 100 x slides 76x26 mm | Holds 20 x slides 76x26 mm |
acrylonitrile-butadiene-styrene (ABS) , white | acrylonitrile-butadiene-styrene (ABS) , white | acrylonitrile-butadiene-styrene (ABS) , black | acrylonitrile-butadiene-styrene (ABS) , white |
pack of 12 ea | pack of 12 ea | pack of 12 ea | pack of 12 ea |
209 mm (8.23 in.) × 172 mm (6.77 in.) × 32 mm (1.26 in.) | 164 mm (6.46 in.) × 110 mm (4.33 in.) × 32 mm (1.26 in.) | 209 mm (8.23 in.) × 172 mm (6.77 in.) × 32 mm (1.26 in.) | 104 mm (4.10 in.) × 94 mm (3.70 in.) × 32 mm (1.26 in.) |
General description
Features and Benefits
- Numbered index sheet in lid for quick identification
- Foam liner
- Stackable
- ABS-durable/impact resistant
- Hinged lid
- Unique sliding compression latch