Documentos clave
Z369047
Finnpipette® Stepper Adapters
non-sterile, For 25/50 mL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
sterility
non-sterile
manufacturer/tradename
Finn
Compatibility
Required for attaching 25 mL and 50 mL tips to Stepper
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries