Documentos clave
P9685
Finnpipette® pipette tips
Finntip 300, non-sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
L:
5.2 cm
maximum volume:
300 μL
volume range:
5-300 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Finn 9401250
material
orange
sterility
non-sterile
feature
barrier: no
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Finn 9401250
L
5.2 cm
maximum volume
300 μL
volume range
5-300 μL
Other Notes
Browse all pipette tips
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)