HS81121B
Inoculating Needle/Loops
loop volume 1 μL, Rigid, sterile, blue
About This Item
Código UNSPSC:
41121800
NACRES:
NB.71
Materiales
blue
esterilidad
sterile
Características
Rigid
envase
pack of 1000 ea
fabricante / nombre comercial
Heathrow Scientific HS81121B
lazo volumen
1 μL
Descripción general
The Inoculating Needle/Loops combination provides choice to spread or pick colonies and cultures. Transfer bacterial cultures without risk of contamination.
Características y beneficios
- Use loop end to uniformly streak a gel surface
- Use needle end for removal of colonies
- Constructed of flexible polypropylene
- Sterile, 20 per zip bag with lot-to-lot traceability
- Disposable
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)