Documentos clave
EPM1282-0600
Base Riser
for Eppendorf® Brunswick Innova® 44/44R Shakers, H 10.2 cm, short
About This Item
UNSPSC Code:
41104405
NACRES:
NB.73
feature
for Eppendorf® Brunswick Innova® 44/44R Shakers
short
manufacturer/tradename
Eppendorf® M1282-0600 (Global)
H
10.2 cm
Categorías relacionadas
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 