Documentos clave
EP5910703007
Adapter for Eppendorf® S-4x400 Rotor
for 13x75 mm-100 mm round-bottom tubes, pkg of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
for 13x75 mm-100 mm round-bottom tubes
packaging
pkg of 2 ea
manufacturer/tradename
Eppendorf® 5910703007
technique(s)
centrifugation: suitable
compatibility
for use with 400 mL round bucket
General description
Adapter for various size tubes and bottles for use with Eppendorf S-4x400 rotor.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG