Documentos clave
EP5820708000
Wide Neck Bottle for Eppendorf® S-4-104 Rotor
round, pack of 2, capacity 750 mL
Sinónimos:
Lid for Eppendorf® FA-45-48-11 Rotor
About This Item
UNSPSC Code:
41121703
NACRES:
NB.43
En este momento no podemos mostrarle ni los precios ni la disponibilidad
material
polypropylene bottle
round
feature
with cap
packaging
pack of 2
manufacturer/tradename
Eppendorf® 5820708000 (Global)
parameter
3250 × g max. RCF
capacity
750 mL
Comparar elementos similares
Ver comparación completa
Mostrar Diferencias
1 of 4
Este artículo
EP022638657EP5820747005EP022638033
manufacturer/tradename
Eppendorf® 5820708000 (Global)
manufacturer/tradename
Eppendorf® 022638657 (US)
manufacturer/tradename
Eppendorf® 5820747005 (Global)
manufacturer/tradename
Eppendorf® 022638033 (NA)
material
polypropylene bottle, round
material
polypropylene bottle, square
material
-
material
-
capacity
750 mL
capacity
500 mL
capacity
-
capacity
-
packaging
pack of 2
packaging
pack of 2
packaging
pack of 2 ea
packaging
pack of 2
parameter
3250 × g max. RCF
parameter
3250 × g max. RCF
parameter
-
parameter
-
Features and Benefits
  • The bottle is made of polypropylene and supplied with a screw cap
  • Compatible for rotors S-4-104, S-4x750, S-4x1000
Legal Information
Eppendorf is a registered trademark of Eppendorf AG