Documentos clave
EP5427762000
Lid for Eppendorf(R) FA-45-48-11 Rotor
aerosol-tight, for 5430/5430R, aluminum
About This Item
UNSPSC Code:
41103913
NACRES:
NB.82
material
aluminum
feature
aerosol-tight
manufacturer/tradename
Eppendorf® 5427762000 (Global)
technique(s)
centrifugation: suitable
compatibility
for use with 5430 R
for use with 5430
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)