Key Documents
EP5409702009
Rotor for Eppendorf® 5427R Centrifuge
FA-45-24-11, aerosol tight, to hold, 24 x 1.5/2.0 mL tubes, lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
lid
to hold 24 x 1.5/2.0 mL tubes
aerosol tight
fixed angle rotor
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5409702009 (Global)
parameter
16,220 rpm
25,000 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5427R
General description
Rotors to fit Eppendorf 5427 R refrigerated centrifuge.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG