Documentos clave
CLS21501L
Pyrex® single cavity chromatography reservoir
capacity 1 L, Corning 2150-1L
Sinónimos:
storage reservoir
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
Pyrex
colorless
grade
for chromatography
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Corning 2150-1L
capacity
1 L
Legal Information
PYREX is a registered trademark of Corning, Inc.