Merck
13471
Sigma-Aldrich
Sulfito de sodio
puriss., meets analytical specification of Ph. Eur., BP, NF, anhydrous, 95.0-100.0% (iodometric)
About This Item
Fórmula lineal:
Na2SO3
Número de CAS:
Peso molecular:
126.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
Quality Level
grade
puriss.
assay
95.0-100.0% (iodometric)
form
solid
quality
meets analytical specification of Ph. Eur., BP, NF
impurities
residual solvents, complies
≤0.001% heavy metals (as Pb)
≤0.05% thiosulfate (S2O3)
pH
9.0-10.5 (25 °C, 126 g/L)
anion traces
chloride (Cl-): ≤200 mg/kg
cation traces
Fe: ≤10 mg/kg
Se: ≤10 mg/kg
Zn: ≤10 mg/kg
suitability
complies for appearance of solution
SMILES string
[Na+].[Na+].[O-]S([O-])=O
InChI
1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2
InChI key
GEHJYWRUCIMESM-UHFFFAOYSA-L
supp_hazards
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Visite la Librería de documentos
Amber R Boles et al.
Biotechnology and bioengineering, 109(3), 637-646 (2011-10-22)
A novel sulfur-utilizing perchlorate reducing bacterial consortium successfully treated perchlorate (ClO₄⁻) in prior batch and bench-scale packed bed reactor (PBR) studies. This study examined the scale up of this process for treatment of water from a ClO ₄⁻ and RDX contaminated
Ben M Goff et al.
Journal of the science of food and agriculture, 92(4), 751-758 (2011-11-19)
A variety of methods have been developed for estimating lignin concentration within plant materials. The objective of this study was to compare the lignin concentrations produced by six methods on a diverse population of forage and biomass materials and to
Yonghong Wu et al.
Analytical biochemistry, 423(1), 15-22 (2012-02-14)
In the current study, we examined the expression level of glyceraldehyde-3-phosphate dehydrogenase (GAPDH) protein in a number of organisms and the stability of GAPDH under various conditions. Our results revealed that GAPDH is present in multiple Escherichia coli strains, the
Qiang Yang et al.
Bioresource technology, 126, 336-340 (2012-11-06)
Agave americana is one of commonly grown agave species but currently less valuable because its large flower stalk cannot be used for producing alcoholic beverage. In the present study, the stalk was pretreated with dilute acid (DA), sulfite (SPORL), and
Michelle Cilia et al.
PloS one, 7(10), e48177-e48177 (2012-11-03)
Circulative transmission of viruses in the Luteoviridae, such as cereal yellow dwarf virus (CYDV), requires a series of precisely orchestrated interactions between virus, plant, and aphid proteins. Natural selection has favored these viruses to be retained in the phloem to