Z742307
Horizontal rotisserie for RotoBot™ Rotating Mixers
holds 6 x 50 mL tubes, pkg of 1 ea
About This Item
Productos recomendados
feature
holds 6 x 50 mL tubes
packaging
pkg of 1 ea
manufacturer/tradename
Benchmark Scientific R4040-HZ50
tube diam.
28-30 mm
General description
For higher capacity applications, these holders can be purchased as accessories for the Rotobot and Rotobot Mini rotating mixers. In addition, horizontal holders are available for applications such as Western blotting and hybridization. The unique, sliding attachment allows for quick and easy changing of tube holders.
Legal Information
Rotobot is a trademark of Benchmark Scientific, Inc.
For use with
Referencia del producto
Descripción
Precios
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)