Z742305
Tube holder for RotoBot™ Rotating Mixers
holds 1 x 50 mL tubes, pack of 2 ea
About This Item
Productos recomendados
feature
holds 1 x 50 mL tubes
packaging
pack of 2 ea
manufacturer/tradename
Benchmark Scientific R4040-500
tube diam.
28-30 mm
Categorías relacionadas
General description
For higher capacity applications, these holders can be purchased as accessories for the Rotobot and Rotobot Mini mixers. Horizontal holders are available for applications such as Western blotting and hybridization. The unique, sliding attachment allows for quick and easy changing of tube holders.
Legal Information
Rotobot is a trademark of Benchmark Scientific, Inc.
For use with
Referencia del producto
Descripción
Precios
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)