Key Documents
Z552445
Sigma-Aldrich
Aldrich® glass thermometer well
fits flask, 2 ‑ 3 L, immersion L 125 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
joint ST/NS24/40
feature
fits flask 2 ‑ 3 L
General description
Knobbed on sides for easy handling.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)