Z531189
Aldrich® coiled condenser
unjacketed, L 500 mm, joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Productos recomendados
joint
joint: ST/NS24/40
L
500 mm
General description
The coiled condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)