Z526193
Glas-Col® Mantleminder II
AC/DC input 120 V AC
About This Item
Productos recomendados
feature
CSA approved
manufacturer/tradename
Glas-Col 104APL512
availability
not available in EU
temperature measuring range
0-750 °C
L
6 in.
General description
- Time proportioning automatic temperature control
- Type J immersion thermocouple included
- Set-point in °C
Legal Information
Glas-Col is a registered trademark of Glas-Col LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)