Z403067
Büchi® receiving flask
capacity 1,000 mL, plastic-coated glass
About This Item
Productos recomendados
material
plastic-coated glass
round bottom
single-neck
joint
joint SJ35/20
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Büchi 20728
capacity
1,000 mL
General description
These fit all Rotavapor models.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)