Key Documents
Z239127
Sigma-Aldrich
White disposable polystyrene pour-boats
W × L × H 86 mm × 128 mm × 25 mm
About This Item
UNSPSC Code:
41100000
NACRES:
NB.71
material
oblong
polystyrene
packaging
pack of 250 ea
manufacturer/tradename
Sigma-Aldrich
W × L × H
86 mm × 128 mm × 25 mm
dish L × W
128 mm × 86 mm
General description
Weighing dish designed to facilitate dispensing and pouring. Smooth uniform walls, flat bottom, usable to 200°F (93°C).
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)