Z189812
Scienceware® industrial and lab funnel
white polyethylene, pkg of 1 ea
About This Item
Productos recomendados
material
white polyethylene
grade
industrial grade
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art H14716-0000
funnel H × O.D.
13 cm × 36 cm
stem H × O.D.
14 cm × 5.1 cm
General description
Molded, 1/4 in. PE funnel provides a greater range of chemical resistance. Fits any 2 in. drum opening.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)