Key Documents
Z127868
Sigma-Aldrich
Aldrich® large separatory funnel
overall capacity 6 L, PTFE stopcock, joint: ST/NS 45/40
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
material
PTFE stopcock
joint
joint: ST/NS45/40
capacity
6 L
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)