Key Documents
SYNT120800
Synthware T-shaped connecting tube
hose connections 8 mm O.D.
Sinónimos:
Synthware T120800, T shape tube connector
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware T120800
General description
Synthware T-shaped connecting tube is designed for use with flexible tubes and/or hoses.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)