PL800GA-NEO-8
Plas-Labs replacement neoprene gloves
for 2 piece glove set, size 8 (for 8 in. ports)
About This Item
UNSPSC Code:
46181504
NACRES:
HA.03
Productos recomendados
material
neoprene
size
8 (for 8 in. ports)
feature
reusable
for use with glove box sleeves
packaging
pack of 1 pair
manufacturer/tradename
Plas-Labs 800GA-NEO-8
General description
Plas-Labs glovebox gloves are available as either one-piece or two-piece assemblies with interchangeable sleeves and gloves. This enables personnel to use the same glove box by changing the size or type of glove being used. Choose from four rubber types. Gloves are 15 mil thickness unless noted otherwise.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
- White Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
- Black neoprene rubber for chemical resistance
- Black butyl rubber for highest impermeability to water vapor, gases, and toxic fluids
- Green nitrile rubber for resistance to oil, fuel, and acids
- Latex rubber for flexibility
Legal Information
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)