Key Documents
GF18433964
Polymethylmethacrylate
(PMMA, ACRYLIC), monofilament, diam. 1 mm, L 10 m, condition optical fiber
Sinónimos:
ME305910
About This Item
UNSPSC Code:
12352100
NACRES:
NA.23
form
monofilament
manufacturer/tradename
Goodfellow 184-339-64
L
10 m
diam.
1 mm
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class
10 - Combustible liquids
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)