Documentos clave
CLS882124
Pyrex® thermometer adapter
joint: ST/NS 24/40
Sinónimos:
lab equipment accessory
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
joint
joint: ST/NS24/40
manufacturer/tradename
Corning 8821-24
L
80 mm
General description
One end with a ground joint and the other end tooled to accommodate a thermometer.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)