BAF249760003
Workshields for β-emitters, free standing
W × L × H 31.4 cm (12.5 in.) × 40.6 cm (16 in.) × 52.1 cm (20.5 in.), size L
About This Item
UNSPSC Code:
26142300
NACRES:
NB.35
Productos recomendados
size
L
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F24976-0003
W × L × H
31.4 cm (12.5 in.) × 40.6 cm (16 in.) × 52.1 cm (20.5 in.)
General description
Select shield size to match requirements.
- 9.5 mm (3/8 in.) thick, glass transparent acrylic shields protect against Beta sources and splashes from lab procedures
- One piece construction has rounded corner for easy cleaning
- Non skid feet prevent slipping on benchtop
- Freestanding design has an angled shield and a flat base with a large work surface
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)