Documentos clave
933287
Sigma-Aldrich
SEM Sample Holder
Multi Pin Holder for 8 Pin Stubs, 3.2mm (1⁄8”)
About This Item
UNSPSC Code:
41111761
NACRES:
NB.35
General description
EMS techware
Application
This 50 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with stainless steel Allen wrench screws. It accommodates up to eight standard 12.7 mm (½") pin stubs.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)