Key Documents
918016
Sigma-Aldrich
Air adaptor
for 3 mL Cartridge
About This Item
UNSPSC Code:
41106401
NACRES:
NA.23
description
Air adaptor, for 3 mL Cartridge, 1 EA
Quality Level
Application
The air adaptor connects cartridge to the air output of a fluid dispenser unit, such as a 3D bioprinter. Each adapter assembly is precision molded for secure attachment to the cartridge with minimal effort, making its connection to the air output of a fluid dispensing unit fast and seamless
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)