Key Documents
58063
Reservoir for Filtration Apparatus 1
volume 250 mL
About This Item
UNSPSC Code:
41115700
volume
250 mL
General description
Replacement glass reservoir for Supelco® mobile phase filtration apparatus 1.
Reservoir for Filtration Apparatus 1(volume 250 mL) protects the instrument and columns by removing particles and gases from solvents and other mobile phase components.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Articles
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.