52782-U
Discovery® DSC-MCAX SPE Tube
bed wt. 100 mg, volume 1 mL, pk of 108
About This Item
Recommended Products
material
PE frit (20 μm)
polypropylene hardware
feature
endcapped
composition
bed wt., 100 mg
packaging
pk of 108
extent of labeling
8.4% C loading
technique(s)
solid phase extraction (SPE): suitable
surface area
480 m2/g
volume
1 mL
matrix
silica gel base material (irregulary shaped, acid washed)
matrix active group
C8 (octyl) bonding
sulfonic acid bonding
particle size
50 μm
pore size
70 Å pore size
capacity
0.06 meq/g capacity
application(s)
food and beverages
separation technique
mixed mode
Looking for similar products? Visit Product Comparison Guide
General description
Sample Matrix Compatibility: Aqueous Solutions (biological fluids, water)
- Packed bed contains both octyl (C8) & benzene sulfonic acid (SCX) bondings
- Dual retention mechanism broadens retention for a range of neutral, basic, acidic and zwitterionic compounds
- Developed for superior selectivity/ sample clean up when isolating basic compounds from biological fluids
- Greater ion-exchange capacity for isolating polar basic and zwitterionic compounds
- Can be used to fractionate basic/ zwitterionic compounds from acidic and neutral compounds
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1A - Skin Sens. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service