Z640549
Eppendorf® epT.I.P.S. Reloads, PCR Clean
volume 500-2500 μL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
500—2500 μL
sterility:
non-sterile
packaging:
pack of 480 (10 trays of 48 tips)
manufacturer/tradename:
Eppendorf® 0030 073.886
Eppendorf® 022491776
Eppendorf® 022491776
Recommended Products
sterility
non-sterile
packaging
pack of 480 (10 trays of 48 tips)
manufacturer/tradename
Eppendorf® 0030 073.886
Eppendorf® 022491776
volume measuring range
500-2500 μL
technique(s)
PCR: suitable
L
115 mm
maximum volume
2.5 mL
volume
500—2500 μL
Looking for similar products? Visit Product Comparison Guide
General description
Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately). PCR clean, free of human DNA, DNase, RNase and PCR inhibitors, batch related certificates available on the internet at http://www.eppendorf.com
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately). PCR clean, free of human DNA, DNase, RNase and PCR inhibitors, batch related certificates available on the internet at http://www.eppendorf.com
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service