Skip to Content
Merck
All Photos(1)

Key Documents

SML1816

Sigma-Aldrich

ABC44

≥98% (HPLC)

Synonym(s):

4-(4-Methoxyphenyl)-1-piperazinecarboxylic acid hexahydro-1,3-dioxo-7-[[1-(phenylmethyl)-4-piperidinyl]methyl]imidazo[1,5-a]pyrazin-2-yl ester, 7-((1-Benzylpiperidin-4-yl)methyl)-1,3-dioxohexahydroimidazo[1,5-a]pyrazin-2(3H)-yl 4-(4-methoxyphenyl)piperazine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H40N6O5
CAS Number:
Molecular Weight:
576.69
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N1OC(N2CCN(C3=CC=C(OC)C=C3)CC2)=O)C(CN(CC4CCN(CC5=CC=CC=C5)CC4)CC6)N6C1=O

InChI

1S/C31H40N6O5/c1-41-27-9-7-26(8-10-27)34-16-18-35(19-17-34)31(40)42-37-29(38)28-23-33(15-20-36(28)30(37)39)22-25-11-13-32(14-12-25)21-24-5-3-2-4-6-24/h2-10,25,28H,11-23H2,1H3

InChI key

YHRQLTHMALGXOG-UHFFFAOYSA-N

Biochem/physiol Actions

ABC44 is a cell-active, potent and selective inhibitor of palmitoyl-protein thioesterase 1 (PPT-1).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Armand B Cognetta et al.
Chemistry & biology, 22(7), 928-937 (2015-06-30)
Serine hydrolase inhibitors, which facilitate enzyme function assignment and are used to treat a range of human disorders, often act by an irreversible mechanism that involves covalent modification of the serine hydrolase catalytic nucleophile. The portion of mammalian serine hydrolases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service