Skip to Content
Merck
All Photos(1)

Key Documents

RF35301715-EP

Eppendorf® Rotor Accessories

steel sleeve, for Rotor F-35-30-17, F-35-30-17, incl. 30 steel sleeves for 15 mL vessels, with 20 adapters for conical tubes and 30 rubber mats for round-bottom tubes, 15 mL

Synonym(s):

Eppendorf® Rotor Accessories

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911

feature

F-35-30-17, incl. 30 steel sleeves for 15 mL vessels, with 20 adapters for conical tubes and 30 rubber mats for round-bottom tubes
Holds 15 mL tubes
steel sleeve, for Rotor F-35-30-17

packaging

pack of 10 ea

manufacturer/tradename

Eppendorf® 022639404
Eppendorf® 022639439

technique(s)

centrifugation: suitable

capacity

15 mL

compatibility

for use with Rotor F-35-30-17

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Eppendorf steel centrifuge tube sleeves designed for use with Eppendorf model F-35-30-17 fixed angle rotors (022639404 or 022639421)
Steel sleeves provide protection for fragile centrifuge tubes during centrifugation

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service