OH30253020
Ohaus® Scout® Portable Balance
Model SPX422, weighing capacity 420 g, resolution: 0.01 g, AC/DC input 110 V, US 3-pin plug (North America)
All Photos(1)
About This Item
UNSPSC Code:
41111502
Recommended Products
material
metal weighing plate
feature
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30253020
weighing capacity measuring range
420 g, resolution: 0.01 g
weighing pan diam.
12 cm
General description
Ideal for laboratory and industrial applications, the OHAUS Scout comes in a slim, stackable design with large backlit LCD. Features include superior overload protection, multiple weighing units and application modes. Geared for high performance in your facility with fast stabilization time and high resolution weighing results, these portable balances set a new standard in laboratory and industrial weighing.
Application
Weighing, Parts Counting, Percent Weighing, Check Weighing, Totalization, Display Hold
Features and Benefits
Stability indicator, stackable design with optional storage cover (excluding 1mg models), mechanical and software overload/underload protection, low battery indicator, auto shut-off, auto tare, span calibration mass included on models up to 620g capacity
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Scout is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)