EPM1289-0500
Test Tube Rack, for Eppendorf® New Brunswick™ Innova® and Eppendorf® Excella® Shakers
size large, holds 22 tubes
All Photos(1)
About This Item
size
large
450x80x129 mm
feature
holds 22 tubes
manufacturer/tradename
Eppendorf® M1289-0500 (Global)
General description
Test Tube Racks have an adjustable angle and tube height support. Made from stainless-steel with inserts to help retain tubes and prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc.
Innova is a registered trademark of Eppendorf, Inc.
New Brunswick is a trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)