Skip to Content
Merck
All Photos(1)

Key Documents

AB-1 HA

12070506, mouse mesothelium, epithelial; small, spindly morphology, cobblestone at confluence

Synonym(s):

AB 1 HA, AB-1HA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

AB-1 HA, 12070506, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

mouse mesothelium

packaging

tube of 5 μg 12070506-DNA-5UG
pkg of vial of cells 12070506-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial; cells are small and can be spindly at low density but form a uniform cobblestone epithelial layer at near confluence

receptors

Not specified

relevant disease(s)

cancer

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Mouse mesothelioma cell line transfected with influenza haemagglutinin gene (HA)

Cell Line Description

AB-1 HA is a mouse mesothelioma cell line transfected with influenza haemagglutinin gene (HA). Mice (BALB/c strain, female, 6-8 weeks old) were exposed to crocidolite asbestos through intraperitoneal injection, resulting in tumour development. Cultures were established from malignant mesothelial cells obtained from ascites fluid. The resulting celll ine, designated AB-1 was transfected with influenza haemagglutinin gene (HA) in order to generate an endogenous tumour antigen. Cell Bank Australia reference number: CBA1374.

DNA Profile

Not specified

Culture Medium

RPMI 1640 (R0883) (with 2 mM L-Glutamine (G7513) + 25mM HEPES) + FBS / FCS (F2442) + 400ug/ml G418.

Subculture Routine

Optimal split ratio 1:10 - 1:20 (seeding density 0.7 x 104 cells/cm2). Cells incubated at 37 °C with 5% CO2. Harvest the cells using 0.05% Trypsin/EDTA at 37 °C for 5 min. Cells will require passaging twice weekly at this seeding density.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments.  Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Richard Lake - University of Western Australia, Australia

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service