Skip to Content
Merck
All Photos(1)

Key Documents

3T3-F442A

70654, mouse adipose tissue, Fibroblast-like

Synonym(s):

3T3F442A Cells, F442A Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

3T3-F442A, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 70654, mouse adipose tissue, Fibroblast-like, 00070654

biological source

mouse adipose tissue

packaging

tube of 5 μg 00070654-DNA-5UG
pkg of vial of cells 00070654-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse pre-adipocytes

Cell Line Description

Cells will differentiate into adipocytes once confluent which takes approximately 10 days. Once confluent, cells should be grown in DMEM and 10% FCS and 5 micrograms/ml insulin. Media changes should take place every 48 h.

To manage customer expectations regarding the potential of 3T3 cell line stocks to differentiate into adipocytes, if using the cells for adipocyte differentiation please note: when cells are stimulated, using an appropriate protocol, differentiation may take several weeks to occur, e.g., 2 - 5 weeks, and the proportion of the population that differentiates can be limited. If 3T3 cells from an alternate source were previously used, we cannot guarantee the differentiation performance will be the same.We are working to source a new stock of this cell line that has a higher rate of adipocyte differentiation potential which we aim to be able to offer in the future. When this is available we will update the cell line details on the website.

Application

Once terminally differentiated the cells can be used as a model for either adipocyte differentiation or mature adipocytes.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3-1:5 ie. seeding at 2-4 x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA, 5% CO2, 37°C. If cells are allowed to become confluent they will differentiate into adipocytes. If cryopreserving these cells, use New

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service