Skip to Content
Merck
All Photos(2)

Key Documents

AV48180

Sigma-Aldrich

Anti-AKR1B1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-ADR, Anti-ALDR1, Anti-ALR2, Anti-AR, Anti-Aldo-keto reductase family 1, member B1 (aldose reductase), Anti-MGC1804

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

pig, mouse, rat, dog, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKR1B1(231)

General description

AKR1B1 codes for a aldo/keto reductase superfamily that catalyzes the reduction of aldehydes. It may functions as PGF synthase in pig endometrium during early pregnancy. AKR1B1 can be induced by proteasome inhibitors in human colon cancer cells. Polymoprhisma in AKR1B1 have been linked to renal insufficiency in type 2 diabetics.
Rabbit Anti-AKR1B1 antibody recognizes chicken, canine, rabbit, human, mouse, rat, bovine, pig, and zebrafish AKR1B1.

Immunogen

Synthetic peptide directed towards the C terminal region of human AKR1B1

Application

Rabbit Anti-AKR1B1 antibody is suitable for western blot applications at a concentration of 1.25μg/ml.

Biochem/physiol Actions

AKR1B1 is a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member catalyzes the reduction of a number of aldehydes, including the aldehyde form of glucose, and is thereby implicated in the development of diabetic complications by catalyzing the reduction of glucose to sorbitol.This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member catalyzes the reduction of a number of aldehydes, including the aldehyde form of glucose, and is thereby implicated in the development of diabetic complications by catalyzing the reduction of glucose to sorbitol. There are a few putative pseudogenes for this gene, and one of them has been confirmed and mapped to chromosome 3. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: QSKGIVVTAYSPLGSPDRPWAKPEDPSLLEDPRIKAIAAKHNKTTAQVLI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heewon Seo et al.
Biology of reproduction, 90(5), 99-99 (2014-04-04)
Prostaglandins (PGs) are important lipid mediators regulating various reproductive processes in many species. In pigs, the expression pattern of PGE2 and PGF2α metabolic enzymes and the regulatory mechanism controlling PGE2 and PGF2α levels in the uterus during pregnancy are not
Nadeeya Mad-Adam et al.
Oncology letters, 23(2), 59-59 (2022-01-08)
Ovarian cancer ranks eighth in cancer incidence and mortality among women worldwide. Cisplatin-based chemotherapy is commonly used for patients with ovarian cancer. However, the clinical efficacy of cisplatin is limited due to the occurrence of adverse side effects and development
Bettina Ebert et al.
Chemico-biological interactions, 191(1-3), 239-249 (2011-01-11)
Aldo-keto reductases (AKRs) play central roles in the reductive metabolism of endogenous signaling molecules and in the detoxification of xenobiotics. AKRC1-1C3, AKR1B1 and AKR1B10 have been shown to be regulated via nuclear factor-erythroid 2 related factor 2 (Nrf2), a transcription
Pushplata Prasad et al.
BMC medical genetics, 11, 52-52 (2010-04-01)
To determine association of nine single nucleotide polymorphisms (SNPs) in ADP ribosyltransferase-1 (ADPRT1), aldo-keto reductase family 1 member B1 (AKR1B1), receptor for advanced glycation end-products (RAGE), glutamine:fructose-6-phosphate amidotransferase-2 (GFPT2), and plasminogen activator inhibitor-1 (PAI-1) genes with chronic renal insufficiency (CRI)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service