Skip to Content
Merck
All Photos(2)

Key Documents

AV41491

Sigma-Aldrich

Anti-GPX3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-GPx-P, Anti-GSHPx-3, Anti-GSHPx-P, Anti-Glutathione Peroxidase 3 (plasma)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
mouse, rat, human, bovine
citations:
technique(s):
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

25 kDa

species reactivity

mouse, rat, human, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPX3(2878)

General description

Glutathione peroxidase refers to a family of isozymes that protect organisms from oxidative damage by reducing lipid hydroperoxides to their corresponding alcohols. Glutathione peroxidase 3 (GPX3) is an extracellular Gpx isozyme found mainly in plasma.

Specificity

Anti-GPX3 polyclonal antibody reacts with human, mouse, rat, bovine, canine, and pig glutathione peroxidase 3 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human GPX3

Application

Anti-GPX3 polyclonal antibody is used to tag glutathione peroxidase-3 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to study the antioxidant lipid protecting role of glutathione peroxidase 3 in extracellular space and plasma.

Biochem/physiol Actions

GPX3 belongs to the glutathione peroxidase family, which functions in the detoxification of hydrogen peroxide. It contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon, which normally signals translation termination.Glutathione peroxidase catalyzes the reduction of hydrogen peroxide, organic hydroperoxide, and lipid peroxides by reduced glutathione and functions in the protection of cells against oxidative damage. Human plasma glutathione peroxidase has been shown to be a selenium-containing enzyme. GPX3 expression appears to be tissue-specific.

Sequence

Synthetic peptide located within the following region: DCHGGISGTIYEYGALTIDGEEYIPFKQYAGKYVLFVNVASYUGLTGQYI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service