Skip to Content
Merck
All Photos(1)

Key Documents

PSRVD01

Multi-Parameter Refractive Index & Density Certified Reference Material at 15 °C, 20 °C and 25 °C, Single 30 mL

Nominal RI Value 1.3330 at 20 °C

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Looking for similar products? Visit Product Comparison Guide

General description

The ISO 17025 / ISO 17034 compliant product provides traceability of measurement to recognized national standards, and to units of measurement realized at the National Physical Laboratory (NPL) or other recognized national standards laboratories. All measurements are fully traceable to National Institute Standards and Technology (NIST).

Application

The combined refractive index and density CRM (nominal RI value 1.3330 at 20 °C) finds its use in the calibration and verification of refractometers such as handheld, Abbe and high-accuracy digital instruments.

Features and Benefits

  • density measured and certified according to ASTM D1480
  • ensures full compliance to ASTM and IP test method protocols
  • low levels of uncertainty, guaranteeing maximum accuracy of data and dependable results
  • supplied in 30 mL volume sealed glass vials for ample measurement readings

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service