Skip to Content
Merck
All Photos(1)

Key Documents

MAB4092

Sigma-Aldrich

Anti-P-Selectin Glycoprotein Ligand-1 Antibody, clone KPL-1

clone KPL-1, Chemicon®, from mouse

Synonym(s):

PSGL-1, CD162

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

KPL-1, monoclonal

species reactivity

human

should not react with

rat (Snapp, KR et al 1998; Baisse, B et al 2007), pig (Snapp, KR et al 1998; Baisse, B et al 2007), bovine (Snapp, KR et al 1998; Baisse, B et al 2007), equine (Snapp, KR et al 1998; Baisse, B et al 2007)

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SELPLG(6404)

General description

Interactions between P-selectin and P-selectin glycoprotein ligand-1 (PSGL-1) mediate the earliest “rolling” of leukocytes on the lumenal surface of endothelial cells at sites of inflammation. To date, the primary role of PSGL-1 in mediating the interaction between neutrophils and P-selectin has been well documented. MAB4092 completely blocks recognition of PSGL-1 by either P-selectin or by L-selectin, but does not affect leukocyte recognition of E-selectin. Two color flow cytometry of normal leukocytes has demonstrated that PSGL-1 is expressed on essentially all blood neutrophils, NK cells, blood cells, T cells, and monocytes, but PSGL-1 stains B cells at significantly lower levels than other cell types. Variation in tyrosine sulfation during B cell differentiation may affect the stability of B cells to interact with P- and L-selectin. {Snapp, KR et al 1998}.

Specificity

P-selectin glycoprotein ligand-1 (PSGL-1). The antibody epitope was mapped to a site within a consensus tyrosine sulfation motif of PSGL-1, previously shown to be essential for interaction with P-selectin (and now shown to be essential for recognition of PSGL-1 by L-selectin). {Snapp KR et al. 1998}. The binding of KPL-1 to human PSGL-1 on HL-60 cells does not require sulfation {Thatte, A et al 2002}.

Immunogen

The antibody epitope was mapped to a site within a consensus tyrosine sulfation motif of PSGL-1, previously shown to be essential for interaction with P-selectin (and now shown to be essential for recognition of PSGL-1 by L-selectin).

Application

Detect P-Selectin Glycoprotein Ligand-1 using this Anti-P-Selectin Glycoprotein Ligand-1 Antibody, clone KPL-1 validated for use in FC, IH, IH(P), IP & WB.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms
Western blot of HL60 whole cell lysates prepared with 1% triton X-100 in 150mM NaCl, 10mM Tris-HCL pH 7.6, 1 mM Ca++, 1 mM Mg++, with 1mmol aprotinin, PMSF,leupeptin, and pepstatin A {Snapp, KR et al 1998}. 1:1000

FACS {Snapp, KR et al 1998}.

Immunoprecipitation: 0.5% triton X-100 in TBS with proteinase inhibitors; 5 µg/500µL of whole cell lysate (350-500mg/mL total protein).

Functional blocking of P-selectin {Snapp KR et al 1998}.

Immunohistochemistry: Fresh frozen, acetone fixed sections

Immunohistochemistry in paraffin embedded tissues: formalin fixed, pretreatment standard citric acid pH 6.0, microwave treatment is recommended 1:50-1:200

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified Immunoglobulin. Liquid in 0.02M PB with 0.25M NaCl, pH7.6, 0.1% Sodium Azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica M Banks et al.
PloS one, 8(11), e78988-e78988 (2013-11-19)
Stem bromelain, a cysteine protease isolated from pineapples, is a natural anti-inflammatory treatment, yet its mechanism of action remains unclear. Curious as to whether bromelain might affect selectin-mediated leukocyte rolling, we studied the ability of bromelain-treated human neutrophils to tether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service