AG30P
Human IgE, myeloma
Synonym(s):
Human Myeloma IgE, IgE Myeloma Protein, IgE from Myeloma
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
human
Quality Level
Assay
≥80% (SDS-PAGE)
manufacturer/tradename
Chemicon®
shipped in
dry ice
target post-translational modification
unmodified
General description
Product Source: Human IgE Myeloma. Source material has been tested for HIV 1, HIV 2, HTLV I, HTLV II, Hepatitis B and C and Syphilis and found to be negative.
Linkage
Replaces: AG30S
Physical form
Format: Purified
Liquid in 10 mM Sodium phosphate, pH 7.2 containing 0.85% sodium chloride and 0.05% sodium azide.
Preparation Note
Purified by deionized water dialysis and gel filtration.
Storage and Stability
Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service