Skip to Content
Merck
All Photos(5)

Key Documents

ABN1661

Sigma-Aldrich

Anti-Membralin

from rabbit

Synonym(s):

Transmembrane protein 259

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
clone:
polyclonal
application:
ICC
IHC
IP
WB
species reactivity:
human, mouse
technique(s):
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
2

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Membralin (UniProt: Q8CIV2; also known as Transmembrane protein 259) is encoded by the Tmem259 (also known as ORF61) gene (Gene ID: 216157) in murine species. Membralin is a highly conserved protein with several transmembrane regions. However, the sequence of membralin does not show any closely related genes. It has multiple glycosylation sites and cytosolic regions at both the N-terminal ends and C-terminal ends. It is expressed in multiple tissues, including brain and spinal cord. However, it is not found in heart, skeletal muscle, and spleen. In humans its membralin expression is reported in certain cancers, including the ovarian cancers. Membralin is reported to have a role in the ER-associated protein degradation (ERAD) pathway and is required for clearance of misfolded proteins in the endoplasmic reticulum (ER). It is also required for survival of motor neurons. Membralin mutant mice display increased basal ER stress and severe and early-onset motor neuron disease and die by postnatal day 5 or 6. (Ref.: Chen YC et al. (2005). Biochimica Biophysica Acta 1730(2): 96-102.)

Specificity

This polyclonal antibody specifically detects membralin in HEK293 cells overexpressing membralin.

Immunogen

A Myc-conjugated linear peptide from the N-terminal region of mouse membralin.
Epitope: N-terminus

Application

Anti-Membralin Antibody, Cat. No. ABN1661, is a highly specific rabbit polyclonal antibody that targets Membralin and has been tested in Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
Immunoprecipitation Analysis: 1.0 µg/mL from a representative lot detected Membralin in lysate from HEK293 cells transfected with mouse membralin. (Courtesy of Dr. Lu-Lin Jiang and Dr. Bing Zhu in Dr. Dongxian Zhang′s Lab at Sanford Burnham Prebys Medical Discovery Institute).

Immunocytochemistry Analysis: 10 µg/mL from a representative lot detected Membralin in mouse neural progenitor cells. (Courtesy of Dr. Lu-Lin Jiang and Dr. Bing Zhu in Dr. Dongxian Zhang′s Lab at Sanford Burnham Prebys Medical Discovery Institute).

Immunohistochemistry Analysis: 5.0 µg/mL from a representative lot detected Membralin in mouse spinal cord tissues. (Courtesy of Dr. Lu-Lin Jiang and Dr. Bing Zhu in Dr. Dongxian Zhang′s Lab at Sanford Burnham Prebys Medical Discovery Institute).

Western Blotting Analysis: 1.0 µg/mL from a representative lot detected Membralin in lysate from HEK293 cells transfected with mouse membralin. (Courtesy of Dr. Lu-Lin Jiang and Dr. Bing Zhu in Dr. Dongxian Zhang′s Lab at Sanford Burnham Prebys Medical Discovery Institute).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in HEK293 cells overexpressing membralin.

Western Blotting Analysis: 1 µg/mL of this antibody detected Membralin in 4 µL of lysate from HEK293 cells transfected with mouse-myc-membralin and overexpressing Membralin.


Target description

~68 kDa observed; 63.56 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein A + Protein G
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bing Zhu et al.
Nature communications, 8(1), 1472-1472 (2017-11-15)
Endoplasmic-reticulum-associated degradation (ERAD) is an important protein quality control system which maintains protein homeostasis. Constituents of the ERAD complex and its role in neurodegeneration are not yet fully understood. Here, using proteomic and FRET analyses, we demonstrate that the ER
Lu-Lin Jiang et al.
The Journal of clinical investigation, 129(8), 3103-3120 (2019-05-22)
Mechanisms underlying motor neuron degeneration in amyotrophic lateral sclerosis (ALS) are yet unclear. Specific deletion of the ER-component membralin in astrocytes manifested postnatal motor defects and lethality in mice, causing the accumulation of extracellular glutamate through reducing the glutamate transporter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service