Skip to Content
Merck

1.04090

SAFC

Glutathione (reduced)

suitable for biopharmaceutical production EMPROVE® bio Ph Eur

Pharma Manufacturing

Synonym(s):

Glutathione (reduced), γ-L-Glutamyl-L-cysteinglycine, GSH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N3O6S
CAS Number:
Molecular Weight:
307.32
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-725-4

Agency

Ph. Eur.

Quality Level

product line

EMPROVE® Bio

pH

3 (20 °C, 10 g/L in H2O)

mp

185-195 °C

solubility

100 g/L

bulk density

160 kg/m3

application(s)

parenterals
parenterals

storage temp.

2-8°C

SMILES string

SCC(NC(=O)CCC([N+H3])C(=O)[O-])C(=O)NCC(=O)O

InChI

1S/C10H17N3O6S/c11-5(10(18)19)1-2-7(14)13-6(4-20)9(17)12-3-8(15)16/h5-6,20H,1-4,11H2,(H,12,17)(H,13,14)(H,15,16)(H,18,19)

InChI key

RWSXRVCMGQZWBV-UHFFFAOYSA-N

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of downstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Analysis Note

Assay (iodometric, calc. on dried substance): 98.0 - 101.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white, slightly agglomerated, crystalline powder
Appearance of solution (100 g/l, water): clear and colorless.
Spec. rotation (α 20/D, 40 g/l, water, calc. on dried substance): -15.5° to -17.5°
Chloride (Cl): ≤ 200 ppm
Sulfate (SO₄): ≤ 300 ppm
As (Arsenic): ≤ 1.5 ppm
Fe (Iron): ≤ 10 ppm
Fe (Iron) (ICP): ≤ 10 ppm
NH₄ (Ammonium): ≤ 200 ppm
Cu (Copper)*: ≤ 25 ppm
Related substances (CE) (Impurity A (Cysteinylglycine)): ≤ 0.5 %
Related substances (CE) (Impurity B (Cysteine)): ≤ 0.5 %
Related substances (CE) (Impurity C (Glutathione oxidized)): ≤ 1.5 %
Related substances (CE) (Impurity D (Glutamylcysteine)): ≤ 1.0 %
Related substances (CE) (Impurity E): ≤ 0.5 %
Related substances (CE) (Largest unspecified impurity): ≤ 0.2 %
Related substances (CE) (Sum of all impurities): ≤ 2.5 %
Acetic acid (HS-GC): ≤ 5000 ppm
Ethanol (HS-GC): ≤ 5000 ppm
Ethyl acetate (HS-GC): ≤ 5000 ppm
Other residual solvents (ICH Q3C): passes test
Sulfated ash (600 °C): ≤ 0.1 %
Loss on drying (105 °C; 3 h): ≤ 0.5 %
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph. Eur.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service