Key Documents
Z743177
Julabo® bath covers
lift-up gable cover to fit CORIO C-B13/17 and CD-B13/17
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9970253
General description
The hinged stainless steel gable hood reduces the evaporation of tempering liquid and saves energy. The rising steam is led back into the bath via a drip edge. This means that the tempering liquid remains in the bath and refill cycles are extended. An additional benefit is that the hood prevents contamination of the samples in the bath. The hinged stainless steel gable hood is designed for operation up to +60°C.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)