WHA68094112
Whatman® Anotop® 25 Plus syringe filter
pore size 0.1 μm, glass microfiber prefilter, sterile
Synonym(s):
Whatman filter, Z747890, syringe filter, syringe filters
About This Item
material
alumina based membrane
polypropylene housing
sterility
sterile
feature
glass microfiber prefilter
packaging
pkg of 50 ea
manufacturer/tradename
Whatman 6809-4112
Whatman Article No. 28420409 (US reference)
diam.
25 mm
volume
≤100 mL
pore size
0.1 μm
fitting
female Luer inlet (lock)
male Luer outlet
Looking for similar products? Visit Product Comparison Guide
Application
- Filtration of tissue culture media
- Clean-up of difficult samples
- Filtration of colloidal material
- Removal of mycoplasma
- HPLC sample preparation
- Biological sample preparation
Features and Benefits
- Contains an inorganic Anopore membrane and is available in three pore sizes
- Polypropylene syringe filters comes in two diameters depending on sample volume
- Integrated glass microfiber prefilter aids in filtration of difficult samples
Suitability
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service