GF39624723
Inconel® alloy 600 - heat resisting alloy, Ni72Cr16Fe8
foil, 150 x 150mm, 0.05mm thickness, annealed, light tight (lt)
Synonym(s):
NI020204
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
description
light tight (lt)
form
annealed (
quarter hard)
foil (
quarter hard)
manufacturer/tradename
Goodfellow 396-247-23
size × thickness
150 mm × 150 mm × 0.05 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service