Skip to Content
Merck
All Photos(1)

Key Documents

BR782082

BRAND® microplate BRANDplates®, cellGrade prem

size 96 wells, size 350 μL, flat bottom, transparent

Synonym(s):

BRANDplates®, Cell culture, Microplates, Tissue culture, cellGrade premium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
flat bottom
polystyrene
transparent wells
size:
350 μL
96 wells
working volume:
350 μL
sterility:
sterile
feature:
lid
skirt (F-bottom)

material

flat bottom
polystyrene
transparent wells

sterility

sterile

feature

lid
skirt (F-bottom)

packaging

pack of 50 ea
pkg of (individually packed)

manufacturer/tradename

BRAND 782082

size

350 μL
96 wells

working volume

350 μL

color

transparent

Looking for similar products? Visit Product Comparison Guide

General description

Poly-D-Lysine-equivalent surface, with analogous results regarding growth performance and cell morphology.Optimal adhesion of cells to the surface reduces cell damage when washing frequently.Cultivation of cell lines with the highest demands on their environment.Surface suited for serum-free and serum-reduced cultivation of cells.Good shelf life at room temperature.Alternative option to biologically coated surfaces.Clearly distinguishable through the color code: orange, embossed alphanumeric coding for 96-well standard plates.Free from endotoxins, DNase, DNA, RNase, non-cytotoxic.PS, sterile, with lids.BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic

Features and Benefits

• Optimal surfaces for optimal results• Reliable quality from the cleanroom• Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service